Learn More
Description
Specifications
Specifications
| Antigen | PSMD8 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Alias | 26S proteasome non-ATPase regulatory subunit 8, 26S proteasome regulatory subunit p31, HIP6,26S proteasome regulatory subunit S14, HYPF, MGC1660, Nin1p, p3126S proteasome regulatory subunit RPN12, proteasome (prosome, macropain) 26S subunit, non-ATPase, 8, Rpn12, S14 |
| Gene Symbols | PSMD8 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
