Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMD9 Polyclonal Antibody

Catalog No. PIPA5121663
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121663 20 μL
PIPA5119364 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121663 Supplier Invitrogen™ Supplier No. PA5121663
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PSMD9 Polyclonal Antibody for Western Blot, IHC (P), Flow

Acts as a chaperone during the assembly of the 26S proteasome, specifically of the base subcomplex of the PA700/19S regulatory complex (RC). During the base subcomplex assembly is part of an intermediate PSMD9:PSMC6:PSMC3 module, also known as modulator trimer complex; PSMD9 is released during the further base assembly process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMD9
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene PSMD9
Gene Accession No. O00233, Q9CR00
Gene Alias 1500011J20Rik; 2610202L11Rik; 26S proteasome non-ATPase regulatory subunit 9; 26S proteasome regulatory subunit p27; Bridge; Bridge-1; homolog of rat Bridge 1; hypothetical protein LOC513315; MGC8644; P27; proteasome (prosome, macropain) 26S subunit, non-ATPase, 9; proteasome 26S non-ATPase regulatory subunit 9; proteasome 26S non-ATPase subunit 9; proteasome 26S subunit, non-ATPase 9; Psmd9; Rpn4; Transactivating protein Bridge-1
Gene Symbols PSMD9
Host Species Rabbit
Immunogen Recombinant protein Proteasome 26S Subunit, Non ATPase 9. The antigen corresponds to amino acid range 2-223 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5715, 67151
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.