Learn More
Invitrogen™ PSMD9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592418
Description
Immunogen sequence: MSDEEARQSG GSSQAGVVTV SDVQELMRRK EEIEAQIKAN YDVLESQKGI GMNEPLVDCE GYPRSDVDLY QVRTARHNII CLQNDHKAVM KQVEEALHQL HARDKEKQAR DMAEAHKEAM SRKLGQSESQ GPPRAFAKVN SISPGSPASI AGLQVDDEIV EFGSVNTQNF QSLHNIGSVV QHSEGKPLNV TVIRRGEKHQ LRLVPTRWAG KGLLGCNIIP LQR; Positive Samples: H460, 22Rv1, Jurkat, A-549.
Acts as a chaperone during the assembly of the 26S proteasome, specifically of the base subcomplex of the PA700/19S regulatory complex (RC). During the base subcomplex assembly is part of an intermediate PSMD9:PSMC6:PSMC3 module, also known as modulator trimer complex; PSMD9 is released during the further base assembly process.
Specifications
| PSMD9 | |
| Polyclonal | |
| Unconjugated | |
| PSMD9 | |
| 1500011J20Rik; 2610202L11Rik; 26S proteasome non-ATPase regulatory subunit 9; 26S proteasome regulatory subunit p27; Bridge; Bridge-1; homolog of rat Bridge 1; hypothetical protein LOC513315; MGC8644; P27; proteasome (prosome, macropain) 26S subunit, non-ATPase, 9; proteasome 26S non-ATPase regulatory subunit 9; proteasome 26S non-ATPase subunit 9; proteasome 26S subunit, non-ATPase 9; Psmd9; Rpn4; Transactivating protein Bridge-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 161475, 5715, 67151 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.79 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00233, Q9CR00, Q9WTV5 | |
| PSMD9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-223 of human PSMD9 (NP_0028042). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.