Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PSMD9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576792
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Acts as a chaperone during the assembly of the 26S proteasome, specifically of the base subcomplex of the PA700/19S regulatory complex (RC). During the base subcomplex assembly is part of an intermediate PSMD9:PSMC6:PSMC3 module, also known as modulator trimer complex; PSMD9 is released during the further base assembly process.
Specifications
| PSMD9 | |
| Polyclonal | |
| Unconjugated | |
| PSMD9 | |
| 1500011J20Rik; 2610202L11Rik; 26S proteasome non-ATPase regulatory subunit 9; 26S proteasome regulatory subunit p27; Bridge; Bridge-1; homolog of rat Bridge 1; hypothetical protein LOC513315; MGC8644; P27; proteasome (prosome, macropain) 26S subunit, non-ATPase, 9; proteasome 26S non-ATPase regulatory subunit 9; proteasome 26S non-ATPase subunit 9; proteasome 26S subunit, non-ATPase 9; Psmd9; Rpn4; Transactivating protein Bridge-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 161475, 5715, 67151 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00233, Q9CR00, Q9WTV5 | |
| PSMD9 | |
| Recombinant full length Human PSMD9. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction