Learn More
PSME2 Polyclonal Antibody, Invitrogen™
Description
PA1-976 detects recombit human and mouse proteasome 11S REG beta. This antibody does not detect endogenous levels of proteasome 11S REG beta nor does it detect other 11S REG subunits. PA1-976 has been successfully used in Western blot and ICC/IF procedures. By Western blot, this antibody detects an ∽28 kDa protein representing recombit human proteasome 11S REG beta. PA1-976 immunizing peptide corresponds to amino acid residues 1-14 from human proteasome 11S REG beta protein. This peptide (PEP-158) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | PSME2 |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Gene | PSME2 |
| Gene Accession No. | Q9UL46, P97372 |
| Gene Alias | PA28B, PA28beta, REGbeta |
| Gene Symbols | PSME2 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.