Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSME4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA11961 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA11961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA11961 Supplier Invitrogen™ Supplier No. PA11961
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

PA1-1961 detects PA200 from bovine, human, mouse and rat samples. PA1-1961 has been successfully used in Western blot, immunofluorescence and immunoprecipitation procedures. By Western blot, this antibody detects an ~200 kDa protein representing PA200 from bovine testis tissue extract. In mouse tissues, this antibody detects both a 200 kDa and a 160 kDa protein. In mouse tissues, the 200 kDa species is most abundant in testis whereas the 160 kDa species is enriched in the spleen. Immunofluorescence staining of PA200 in HeLa cells results in diffuse nuclear staining prior to ionizing radiation treatment. Immunofluorescence staining of PA200 in HeLa cells after ionizing radiation treatment results in focal aggregate staining within the nucleus.

Associated component of the proteasome that specifically recognizes acetylated histones and promotes ATP- and ubiquitin-independent degradation of core histones during spermatogenesis and DNA damage response. Recognizes and binds acetylated histones via its bromodomain-like (BRDL) region and activates the proteasome by opening the gated channel for substrate entry. Binds to the core proteasome via its C-terminus, which occupies the same binding sites as the proteasomal ATPases, opening the closed structure of the proteasome via an active gating mechanism. Component of the spermatoproteasome, a form of the proteasome specifically found in testis: binds to acetylated histones and promotes degradation of histones, thereby participating actively to the exchange of histones during spermatogenesis. Also involved in DNA damage response in somatic cells, by promoting degradation of histones following DNA double-strand breaks. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSME4
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.05% sodium azide
Gene PSME4
Gene Accession No. F1MKX4, Q14997, Q5SSW2
Gene Alias AA409398; AU041366; Kiaa0077; mKIAA0077; PA200; proteasome (prosome, macropain) activator subunit 4; proteasome activator 200 kDa; proteasome activator complex subunit 4; Proteasome activator PA200; proteasome activator subunit 4; Protein TEMO; Psme4; TEMO
Gene Symbols PSME4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues TKLPKKRKRDPGSVG of human PA200 (aa 1734-1748).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103554, 23198, 498433, 528142
Target Species Human, Mouse, Rat, Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.