Learn More
Invitrogen™ PSME4 Polyclonal Antibody

Description
Positive Samples: MCF-7, U-87MG, 293T, HeLa, Mouse thymus, Mouse brain, Mouse heart, Mouse kidney, Rat liver; Cellular Location: Cytoplasm, Nucleus, Nucleus speckle, cytosol.
Specifications
Specifications
| Antigen | PSME4 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.38 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | PSME4 |
| Gene Accession No. | Q14997, Q5SSW2 |
| Gene Alias | AA409398; AU041366; Kiaa0077; mKIAA0077; PA200; proteasome (prosome, macropain) activator subunit 4; proteasome activator 200 kDa; proteasome activator complex subunit 4; Proteasome activator PA200; proteasome activator subunit 4; Protein TEMO; Psme4; TEMO |
| Gene Symbols | PSME4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.