Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PSME4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5141041
This item is not returnable.
View return policy
Description
PSME4 Polyclonal Antibody for Western Blot, IHC (P)
Associated component of the proteasome that specifically recognizes acetylated histones and promotes ATP- and ubiquitin-independent degradation of core histones during spermatogenesis and DNA damage response. Recognizes and binds acetylated histones via its bromodomain-like (BRDL) region and activates the proteasome by opening the gated channel for substrate entry. Binds to the core proteasome via its C-terminus, which occupies the same binding sites as the proteasomal ATPases, opening the closed structure of the proteasome via an active gating mechanism. Component of the spermatoproteasome, a form of the proteasome specifically found in testis: binds to acetylated histones and promotes degradation of histones, thereby participating actively to the exchange of histones during spermatogenesis. Also involved in DNA damage response in somatic cells, by promoting degradation of histones following DNA double-strand breaks. [UniProt].
Specifications
| PSME4 | |
| Polyclonal | |
| Unconjugated | |
| PSME4 | |
| AA409398; AU041366; Kiaa0077; mKIAA0077; PA200; proteasome (prosome, macropain) activator subunit 4; proteasome activator 200 kDa; proteasome activator complex subunit 4; Proteasome activator PA200; proteasome activator subunit 4; Protein TEMO; Psme4; TEMO | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 103554, 23198, 498433 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q14997, Q5SSW2 | |
| PSME4 | |
| Recombinant protein within human PSME4 aa 1,550-1,843. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction