Learn More
PSME4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PSME4 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2228
![]() ![]() |
Proteintech Group Inc
187991AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody is expected to recognize all 4 isoforms of PSME4 but appears to mainly detect isoform 2 of 140 kDa.
proteasome (prosome, macropain) activator subunit 4 (PSME4) is subunit of the proteasome. PSME4 has been found to activate proteasomal cleavage of peptides in an energy-independent manner and may be involved in spermatogenesis. PSME4 may also be involved in DNA repair.Specifications
PSME4 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q14997, Q5SSW2 | |
103554, 23198, 498433 | |
Peptide | |
Primary | |
-20°C | |
PSME4 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA0077, PA200, Proteasome activator PA200, PSME4 | |
Psme4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.