Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMF1 Antibody - Isoform 1, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB1001178
Description
PSMF1 Polyclonal specifically detects PSMF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Peptide ELISA, Flow (Intracellular).Specifications
PSMF1 Isoform 1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
PI31, proteasome (prosome, macropain) inhibitor subunit 1, proteasome (prosome, macropain) inhibitor subunit 1 (PI31), PSMF1 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize isoform 1 (NP_006805.2 and NP_848693.2). | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Flow Cytometry | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000, Flow (Intracellular) | |
NP_006805 | |
PSMF1 | |
Peptide with sequence C-DHLPPPGYDDMYL corresponding to C-Terminus according to NP_006805.2. | |
0.1 mg | |
Proteases & Other Enzymes | |
9491 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction