Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMF1 Antibody (OTI2D7), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273707AF594
Description
PSMF1 Monoclonal specifically detects PSMF1 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PSMF1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
PSMF1 | |
Full length recombinant protein of human PSMF1 (NP_848693) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2D7 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
hPI31, PI31, proteasome (prosome, macropain) inhibitor subunit 1 (PI31), proteasome inhibitor hP131 subunit, proteasome inhibitor PI31 subunit | |
Mouse | |
Affinity Purified | |
Proteases & Other Enzymes | |
9491.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction