Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMF1 Antibody (OTI2D7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245903
Description
Ensure accurate, reproducible results in Western Blot, Immunofluorescence
PSMF1 Monoclonal specifically detects PSMF1 in Human, Rat samples. It is validated for Western Blot, Immunofluorescence.Specifications
PSMF1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:200 - 1:2000, Immunofluorescence 1:100 | |
hPI31, PI31, proteasome (prosome, macropain) inhibitor subunit 1 (PI31), proteasome inhibitor hP131 subunit, proteasome inhibitor PI31 subunit | |
Mouse | |
29.6 kDa | |
0.1 mL | |
Proteases & Other Enzymes | |
9491 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunofluorescence | |
OTI2D7 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
PSMF1 | |
Full length human recombinant protein of human PSMF1(NP_848693) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction