Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PSMG1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519076
Description
This antibody is predicted to react with bovine, canine, mouse, rabbit and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
Chaperone protein which promotes assembly of the 20S proteasome as part of a heterodimer with PSMG2. The PSMG1-PSMG2 heterodimer binds to the PSMA5 and PSMA7 proteasome subunits, promotes assembly of the proteasome alpha subunits into the heteroheptameric alpha ring and prevents alpha ring dimerization.
Specifications
PSMG1 | |
Polyclonal | |
Unconjugated | |
PSMG1 | |
AW552102; C21LRP; C21-LRP; Chromosome 21 leucine-rich protein; Down syndrome critical region gene 2; Down syndrome critical region homolog 2; Down syndrome critical region protein 2; Down syndrome critical region protein 2 homolog; Dscr2; leucine rich protein C21-LRP; LRPC21; PAC1; PAC-1; proteasome (prosome, macropain) assembly chaperone 1; proteasome assembling chaperone 1; proteasome assembly chaperone 1; Psmg1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
8624 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O95456 | |
PSMG1 | |
Synthetic peptide sequence (KLMTTNEIQSNIYT) corresponding to the C-terminus amino acids of PSMG1 (aa 275-288). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction