Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PSMG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559610
Description
Immunogen sequence: GFTLLMPAVS VGNVGQLAMD LIISTLNMSK IGYFYTDCLV PMVGNNPYAT TEGNSTELSI NAEVYSLPSR KLVALQ Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.
Chaperone protein which promotes assembly of the 20S proteasome as part of a heterodimer with PSMG1. The PSMG1-PSMG2 heterodimer binds to the PSMA5 and PSMA7 proteasome subunits, promotes assembly of the proteasome alpha subunits into the heteroheptameric alpha ring and prevents alpha ring dimerization.
Specifications
PSMG2 | |
Polyclonal | |
Unconjugated | |
Psmg2 | |
1700017I17Rik; AW545363; CD40 ligand-activated specific transcript 3; CLAST3; HCCA3; HDCMC29P; hepatocellular carcinoma susceptibility protein; hepatocellular carcinoma-susceptibility protein 3; HSPC260; HsT1707; likely ortholog of mouse CD40 ligand-activated specific transcript 3 (Clast3); MDS003; PAC2; PAC-2; proteasome (prosome, macropain) assembly chaperone 2; proteasome a; proteasome assembling chaperone 2; proteasome assembly chaperone 2; PSMG2; TNFSF5IP1; Tumor necrosis factor superfamily member 5-induced protein 1; tumor necrosis factor superfamily, member 5-induced protein 1; x 003 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56984 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969U7 | |
Psmg2 | |
Recombinant protein corresponding to Human PSMG2. Recombinant protein control fragment (Product #RP-98232). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction