Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561301
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561301 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561301 Supplier Invitrogen™ Supplier No. PA561301
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IPGGGITKTL YDESCSKEIQ MAVLLKFVSE GDNIPDALGL VEYLNEWLQI LKPLSDDPTV SASRWKIPSS WRLLFGSGLP Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.

Chaperone protein which promotes assembly of the 20S proteasome as part of a heterodimer with PSMG1. The PSMG1-PSMG2 heterodimer binds to the PSMA5 and PSMA7 proteasome subunits, promotes assembly of the proteasome alpha subunits into the heteroheptameric alpha ring and prevents alpha ring dimerization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMG2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Psmg2
Gene Accession No. Q969U7
Gene Alias 1700017I17Rik; AW545363; CD40 ligand-activated specific transcript 3; CLAST3; HCCA3; HDCMC29P; hepatocellular carcinoma susceptibility protein; hepatocellular carcinoma-susceptibility protein 3; HSPC260; HsT1707; likely ortholog of mouse CD40 ligand-activated specific transcript 3 (Clast3); MDS003; PAC2; PAC-2; proteasome (prosome, macropain) assembly chaperone 2; proteasome a; proteasome assembling chaperone 2; proteasome assembly chaperone 2; PSMG2; TNFSF5IP1; Tumor necrosis factor superfamily member 5-induced protein 1; tumor necrosis factor superfamily, member 5-induced protein 1; x 003 protein
Gene Symbols Psmg2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PSMG2. Recombinant protein control fragment (Product #RP-98231).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56984
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.