Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104392
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104392 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104392 Supplier Invitrogen™ Supplier No. PA5104392
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PSMG2.

Chaperone protein which promotes assembly of the 20S proteasome as part of a heterodimer with PSMG1. The PSMG1-PSMG2 heterodimer binds to the PSMA5 and PSMA7 proteasome subunits, promotes assembly of the proteasome alpha subunits into the heteroheptameric alpha ring and prevents alpha ring dimerization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMG2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Psmg2
Gene Accession No. Q969U7, Q9EST4
Gene Alias 1700017I17Rik; AW545363; CD40 ligand-activated specific transcript 3; CLAST3; HCCA3; HDCMC29P; hepatocellular carcinoma susceptibility protein; hepatocellular carcinoma-susceptibility protein 3; HSPC260; HsT1707; likely ortholog of mouse CD40 ligand-activated specific transcript 3 (Clast3); MDS003; PAC2; PAC-2; proteasome (prosome, macropain) assembly chaperone 2; proteasome a; proteasome assembling chaperone 2; proteasome assembly chaperone 2; PSMG2; TNFSF5IP1; Tumor necrosis factor superfamily member 5-induced protein 1; tumor necrosis factor superfamily, member 5-induced protein 1; x 003 protein
Gene Symbols Psmg2
Host Species Rabbit
Immunogen A synthesized peptide derived from human PSMG2(Accession Q969U7), corresponding to amino acid residues E104-K154.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107047, 291539, 56984
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.