Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMG3 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297973R
Description
PSMG3 Polyclonal specifically detects PSMG3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PSMG3 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C7orf48, hPAC3, MGC10911, PAC-3, PAC3chromosome 7 open reading frame 48, proteasome (prosome, macropain) assembly chaperone 3, proteasome assembling chaperone 3, proteasome assembly chaperone 3 | |
Produced in rabbits immunized with E. coli-derived Human PSMG3 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
84262 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction