Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114157
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114157 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114157 Supplier Invitrogen™ Supplier No. PA5114157
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLLGQDEPLI HVFAKNLVAF VSQEAGNRAV LLAVAVKDKS MEGLKALREV For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 77%; Guinea Pig: 93%; Horse: 85%; Human: 100%; Mouse: 86%; Rat: 93%.

PSMG3, also named as C7orf48 and PAC3, is a chaperone protein which promotes assembly of the 20S proteasome. It may cooperate with PSMG1-PSMG2 heterodimers to orchestrate the correct assembly of proteasomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMG3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Psmg3
Gene Accession No. Q9BT73
Gene Alias 1810042K04Rik; 4930403H09Rik; AI303239; C7orf48; hPAC3; mPAC3; PAC3; PAC-3; proteasome (prosome, macropain) assembly chaperone 3; proteasome (prosome, macropain) assembly chaperone 3-like; proteasome assembling chaperone 3; proteasome assembly chaperone 3; Psmg3
Gene Symbols Psmg3
Host Species Rabbit
Immunogen Immunogen sequence: VLLGQDEPLI HVFAKNLVAF VSQEAGNRAV LLAVAVKDKS MEGLKALREV (aa 66-115).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84262
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.