Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMG4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140040
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140040 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140040 Supplier Invitrogen™ Supplier No. PA5140040
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LQNTDSNFAL LVENRIKEEM EAFPEKF Highest antigen sequence indentity to the following orthologs: Mouse - 89%, Rat - 89%.

Chaperone protein which promotes assembly of the 20S proteasome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMG4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PSMG4
Gene Accession No. Q5JS54
Gene Alias bA506K6.2; C6orf86; hPAC4; PAC4; PAC-4; proteasome (prosome, macropain) assembly chaperone 4; proteasome assembly chaperone 4; PSMG4
Gene Symbols PSMG4
Host Species Rabbit
Immunogen Recombinant protein fragment of Human PSMG4 (Product #RP-107593)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389362
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.