Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Psora 4
GREENER_CHOICE

Catalog No. 436710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-671-0 10 mg
43-675-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-671-0 Supplier Tocris Bioscience™ Supplier No. 4367/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent KV1.3 channel blocker

Potent KV1.3 channel blocker (EC50 = 3 nM). Preferentially binds the C-type inactivated state of the channel, similar to the activity of CP 339818 (Cat. No. 1399). Displays 17- to 70-fold selectivity over closely-related KV1 channels (KV1, KV2, KV4 and KV7); exhibits activity at KV1.5 (IC50 = 7.7 nM). Suppresses proliferation of human and rat myelin-specific effector memory T cells (EC50 values are 25 and 60 nM respectively).

Chemical Identifiers

CAS 724709-68-6
Molecular Formula C21H18O4

Specifications

Quantity 10 mg
Formula Weight 334.37
Purity >99%
Molecular Weight (g/mol) 334.371
Chemical Name or Material Psora 4
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.