Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSP/BPIFA2 Antibody (1F12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP237493
Description
PSP/BPIFA2 Monoclonal specifically detects PSP/BPIFA2 in Human, Mouse, Porcine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
PSP/BPIFA2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
bA49G10.1, chromosome 20 open reading frame 70, Parotid secretory protein, PSP, short palate, lung and nasal epithelium carcinoma associated 2, short palate, lung and nasal epithelium carcinoma-associated protein 2, SPLUNC2 | |
Mouse | |
Affinity Purified | |
RUO | |
140683 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1F12 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
BPIFA2 | |
Purified recombinant fragment of mouse Splunc2 expressed in E. Coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Pig | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction