Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSP/BPIFA2 Antibody (5B2), Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | PSP/BPIFA2 |
---|---|
Clone | 5B2 |
Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 |
Applications | Western Blot, Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
PSP/BPIFA2 Monoclonal specifically detects PSP/BPIFA2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
PSP/BPIFA2 | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
Monoclonal | |
Ascites | |
RUO | |
Ascites with 0.03% Sodium Azide | |
140683 | |
Purified recombinant fragment of human Splunc2 expressed in E. Coli. | |
Primary | |
27 kDa |
5B2 | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
Human | |
bA49G10.1, chromosome 20 open reading frame 70, Parotid secretory protein, PSP, short palate, lung and nasal epithelium carcinoma associated 2, short palate, lung and nasal epithelium carcinoma-associated protein 2, SPLUNC2 | |
BPIFA2 | |
IgG1 | |
Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title