Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSP (REG1A) Monoclonal Antibody (OTI8D5), TrueMAB™, OriGene
SDP

Catalog No. 502543515
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3515 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3515 Supplier OriGene Supplier No. CF813969
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSP (REG1A) Monoclonal antibody specifically detects PSP (REG1A) in Human samples. It is validated for ELISA

Specifications

Antigen PSP (REG1A)
Applications ELISA
Classification Monoclonal
Clone OTI8D5
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene REG1A
Gene Accession No. P05451
Gene Alias ICRF; islet cells regeneration factor; Islet of Langerhans regenerating protein; LITHOST; Lithostathine; lithostathine 1 alpha; lithostathine-1-alpha; MGC12447; P19; pancreatic stone protein; pancreatic stone protein, secretory; pancreatic thread protein; protein-X; PSP; PSPS; PSPS1; PTP; RATLITHOST; RATRGPI; Reg; Reg1; REG1A; REG-1-alpha; regenerating family member 1 alpha; regenerating islet-derived 1 alpha; regenerating islet-derived mouse homolog 1; regenerating islet-derived protein 1-alpha; Regenerating protein I alpha; regeneration protein, lithostatin, pancreatic stone protein; Rgp1; RGPI
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 23-166 of human PSP (NP_002900) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5967
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.