Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSP94/MSMB Antibody (OTI3D5), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273625APC
Description
PSP94/MSMB Monoclonal antibody specifically detects PSP94/MSMB in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
PSP94/MSMB | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
OTI3D5 | |
Western Blot, ELISA, Immunohistochemistry | |
IGBF, MSMB, MSPB, PIP, PN44, PRPS, PSP57, PSP94 | |
Purified prostate specific antigen (MSMB) from human seminal fluid | |
0.1 mL | |
Cancer | |
4477 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction