Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSP94/MSMB Antibody (OTI6C7), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273624F
Description
PSP94/MSMB Monoclonal specifically detects PSP94/MSMB in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| PSP94/MSMB | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| MSMB | |
| Full length recombinant protein of human Prostate Secretory Protein/PSP (NP_002434) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI6C7 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| IGBF, MSMB, MSPB, PIP, PN44, PRPS, PSP57, PSP94 | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 4477.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction