Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSPH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554189
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554189 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554189 Supplier Invitrogen™ Supplier No. PA554189
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AVCFDVDSTV IREEGIDELA KICGVEDAVS EMTRRAMGGA VPFKAALTER LALIQPSREQ VQRLIAEQPP HLTPGIRELV SRLQERNVQV FLISG Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

The protein encoded by this gene belongs to a subfamily of the phosphotransferases. This encoded enzyme is responsible for the third and last step in L-serine formation. It catalyzes magnesium-dependent hydrolysis of L-phosphoserine and is also involved in an exchange reaction between L-serine and L-phosphoserine. Deficiency of this protein is thought to be linked to Williams syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSPH
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PSPH
Gene Accession No. P78330
Gene Alias AI480570; L-3-phosphoserine phosphatase; O-phosphoserine phosphohydrolase; OTTHUMP00000025059; Phosphoserine phosphatase; PSP; PSPase; Psph; PSPHD
Gene Symbols PSPH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PSPH. Recombinant protein control fragment (Product #RP-92746).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5723
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.