Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSPH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596863
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596863 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596863 Supplier Invitrogen™ Supplier No. PA596863
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVSHSELRKL FYSADAVCFD VDSTVIREEG IDELAKICGV EDAVSEMTRR AMGGAVPFKA ALTERLALIQ PSREQVQRLI AEQPPHLTPG IRELVSRLQE RNVQVFLISG GFRSIVEHVA SKLNIPATNV FANRLKFYFN GEYAGFDETQ PTAESGGKGK VIKLLKEKFH FKKIIMIGDG ATDMEACPPA DAFIGFGGNV IRQQVKDNAK WYITDFVELL GELEE; Positive Samples: HeLa, HepG2, U-251MG, Jurkat, A-431, Mouse brain, Mouse kidney, Mouse testis, Rat liver.

The protein encoded by this gene belongs to a subfamily of the phosphotransferases. This encoded enzyme is responsible for the third and last step in L-serine formation. It catalyzes magnesium-dependent hydrolysis of L-phosphoserine and is also involved in an exchange reaction between L-serine and L-phosphoserine. Deficiency of this protein is thought to be linked to Williams syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSPH
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PSPH
Gene Accession No. P78330, Q5M819, Q99LS3
Gene Alias AI480570; L-3-phosphoserine phosphatase; O-phosphoserine phosphohydrolase; OTTHUMP00000025059; Phosphoserine phosphatase; PSP; PSPase; Psph; PSPHD
Gene Symbols PSPH
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-225 of human PSPH (NP_0045682).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100678, 304429, 5723
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.