Learn More
Invitrogen™ PSRC1 Monoclonal Antibody (GT982)

Description
Positive Control: human PSRC1-transfected 293T cells Predicted Reactivity: Rhesus Monkey (95%), Bovine (86%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | PSRC1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | GT982 |
| Concentration | 1.37 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | PSRC1 |
| Gene Accession No. | Q6PGN9 |
| Gene Alias | 5430413I02Rik; AL033312; Dda3; differential display and activated by p53; FP3214; MGC1780; OTTHUMP00000013785; OTTHUMP00000013786; p53-regulated DDA3; proline and serine rich coiled-coil 1; proline/serine-rich coiled-coil 1; proline/serine-rich coiled-coil protein 1; Psrc1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.