Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSRC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566023
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566023 Supplier Invitrogen™ Supplier No. PA566023
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NRLAAQLEQCA LQDRESAGEG LGPRRVKPSP RRETFVLKDS PVRDLLPTVN SLTRSTPSPS SLTPRLRSND RKGSVRALRA TSGKRPSNMK R Highest antigen sequence identity to the following orthologs - mouse 70%, rat 67%.

PSRC1 is required for normal progression through mitosis. Required for normal congress of chromosomes at the metaphase plate, and for normal rate of chromosomal segregation during anaphase. Plays a role in the regulation of mitotic spindle dynamics. Increases the rate of turnover of microtubules on metaphase spindles, and contributes to the generation of normal tension across sister kinetochores. Recruits KIF2A and ANKRD53 to the mitotic spindle and spindle poles. May participate in p53/TP53-regulated growth suppression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSRC1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PSRC1
Gene Accession No. Q6PGN9
Gene Alias 5430413I02Rik; AL033312; Dda3; differential display and activated by p53; FP3214; MGC1780; OTTHUMP00000013785; OTTHUMP00000013786; p53-regulated DDA3; proline and serine rich coiled-coil 1; proline/serine-rich coiled-coil 1; proline/serine-rich coiled-coil protein 1; Psrc1
Gene Symbols PSRC1
Host Species Rabbit
Immunogen Recombinant Human PSRC1. Recombinant protein control fragment (Product #RP-106713).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84722
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.