Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSRC1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA578606

Catalog No. PIPA578606


Only null left
Add to Cart

Description

Description

Positive Control: 293T, A431, human PSRC1-transfected 293T Predicted Reactivity: Rhesus Monkey (95%), Bovine (86%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PSRC1 is required for normal progression through mitosis. Required for normal congress of chromosomes at the metaphase plate, and for normal rate of chromosomal segregation during anaphase. Plays a role in the regulation of mitotic spindle dynamics. Increases the rate of turnover of microtubules on metaphase spindles, and contributes to the generation of normal tension across sister kinetochores. Recruits KIF2A and ANKRD53 to the mitotic spindle and spindle poles. May participate in p53/TP53-regulated growth suppression.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PSRC1
Polyclonal
Unconjugated
PSRC1
5430413I02Rik; AL033312; Dda3; differential display and activated by p53; FP3214; MGC1780; OTTHUMP00000013785; OTTHUMP00000013786; p53-regulated DDA3; proline and serine rich coiled-coil 1; proline/serine-rich coiled-coil 1; proline/serine-rich coiled-coil protein 1; Psrc1
Rabbit
Antigen Affinity Chromatography
RUO
56742, 84722
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.86 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
Q6PGN9, Q9D0P7
PSRC1
Recombinant protein encompassing a sequence within the center region of human PSRC1. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.