Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSRC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596339
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596339 Supplier Invitrogen™ Supplier No. PA596339
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEDLEEDVRF IVDETLDFGG LSPSDSREEE DITVLVTPEK PLRRGLSHRS DPNAVAPAPQ GVRLSLGPLS PEKLEEILDE ANRLAAQLEQ CALQDRESAG EGLGPRRVKP SPRRETFVLK DSPVRDLLPT VNSLTRSTPS PSSLTPRLRS NDRKGSVRAL RATSGKRPSN MKRESPTCNL FPASKSPASS PLTRSTPPVR GRAGPSGRAA ASEET; Positive Samples: MCF7, HepG2, 293T; Cellular Location: Cytoplasm, cytoskeleton, spindle, spindle pole.

PSRC1 is required for normal progression through mitosis. Required for normal congress of chromosomes at the metaphase plate, and for normal rate of chromosomal segregation during anaphase. Plays a role in the regulation of mitotic spindle dynamics. Increases the rate of turnover of microtubules on metaphase spindles, and contributes to the generation of normal tension across sister kinetochores. Recruits KIF2A and ANKRD53 to the mitotic spindle and spindle poles. May participate in p53/TP53-regulated growth suppression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSRC1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PSRC1
Gene Accession No. Q6PGN9
Gene Alias 5430413I02Rik; AL033312; Dda3; differential display and activated by p53; FP3214; MGC1780; OTTHUMP00000013785; OTTHUMP00000013786; p53-regulated DDA3; proline and serine rich coiled-coil 1; proline/serine-rich coiled-coil 1; proline/serine-rich coiled-coil protein 1; Psrc1
Gene Symbols PSRC1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-215 of human PSRC1 (NP_0010052901).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84722
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.