Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSTK Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | PSTK |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP170690
![]() |
Novus Biologicals
NBP170690 |
100 μL |
Each for $487.50
|
|
|||||
NBP17069020
![]() |
Novus Biologicals
NBP17069020UL |
20 μL | N/A | N/A | N/A | ||||
Description
PSTK Polyclonal specifically detects PSTK in Human samples. It is validated for Western Blot.Specifications
| PSTK | |
| Polyclonal | |
| Rabbit | |
| phosphoseryl-tRNA kinase | |
| PSTK | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 118672 | |
| Synthetic peptides corresponding to PSTK(phosphoseryl-tRNA kinase) The peptide sequence was selected from the middle region of PSTK. Peptide sequence SRPLFLVLDDNFYYQSMRYEVYQLARKYSLGFCQLFLDCPLETCLQRNGQ. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title