Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PSTK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558100
Description
Immunogen sequence: KTAENIRGTG SDGPRKRGLC VLCGLPAAGK STFARALAHR LQQEQGWAIG VVAYDDVMPD AFLAGARARP APSQWKLLRQ E Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 65%.
PSTK specifically phosphorylates seryl-tRNA(Sec) to O-phosphoseryl-tRNA(Sec), an activated intermediate for selenocysteine biosynthesis.
Specifications
PSTK | |
Polyclonal | |
Unconjugated | |
PSTK | |
C10orf89; L-seryl-tRNA(Sec) kinase; O-phosphoseryl-tRNA(Sec) kinase; phosphoseryl-tRNA kinase; PSTK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
118672 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IV42 | |
PSTK | |
Recombinant protein corresponding to Human PSTK. Recombinant protein control fragment (Product #RP-103992). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction