Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSTPIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578276
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578276 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578276 Supplier Invitrogen™ Supplier No. PA578276
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted Reactivity: Mouse (83%), Rat (80%), Pig (85%), Bovine (86%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PSTPIP1 is involved in regulation of the actin cytoskeleton. May regulate WAS actin-bundling activity. Bridges the interaction between ABL1 and PTPN18 leading to ABL1 dephosphorylation. May play a role as a scaffold protein between PTPN12 and WAS and allow PTPN12 to dephosphorylate WAS. Has the potential to physically couple CD2 and CD2AP to WAS. Acts downstream of CD2 and CD2AP to recruit WAS to the T-cell:APC contact site so as to promote the actin polymerization required for synapse induction during T-cell activation. Down-regulates CD2-stimulated adhesion through the coupling of PTPN12 to CD2. Also has a role in innate immunity and the inflammatory response. Recruited to inflammasomes by MEFV. Induces formation of pyroptosomes, large supramolecular structures composed of oligomerized PYCARD dimers which form prior to inflammatory apoptosis. Binding to MEFV allows MEFV to bind to PYCARD and facilitates pyroptosome formation. Regulates endocytosis and cell migration in neutrophils.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSTPIP1
Applications Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene PSTPIP1
Gene Accession No. O43586
Gene Alias CD2 antigen-binding protein 1; CD2 cytoplasmic tail-binding protein; CD2-binding protein 1; CD2BP1; CD2BP1L; CD2BP1S; def-2; H-PIP; PAPAS; PEST phosphatase-interacting protein 1; proline-serine-threonine phosphatase interacting protein 1; proline-serine-threonine phosphatase-interacting protein 1; PSTPIP; Pstpip1; truncated proline-serine-threonine phosphatase interacting protein 1
Gene Symbols PSTPIP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human PSTPIP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9051
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.