Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSTPIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528913
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528913 Supplier Invitrogen™ Supplier No. PA528913
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Raw264.7. Predicted reactivity: Mouse (88%), Rat (86%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Binds to F-actin. May be involved in regulation of the actin cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSTPIP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene PSTPIP2
Gene Accession No. Q99M15, Q9H939
Gene Alias cmo; Macrophage actin-associated tyrosine-phosphorylated protein; MAYP; PEST phosphatase-interacting protein 2; pp37; proline-serine-threonine phosphatase interacting protein 2; proline-serine-threonine phosphatase-interacting protein 2; PSTPIP2; RGD1563090
Gene Symbols PSTPIP2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PSTPIP2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19201, 9050
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.