Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSTPIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598992
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598992 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598992 Supplier Invitrogen™ Supplier No. PA598992
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PSTPIP2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Binds to F-actin. May be involved in regulation of the actin cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSTPIP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PSTPIP2
Gene Accession No. Q9H939
Gene Alias cmo; Macrophage actin-associated tyrosine-phosphorylated protein; MAYP; PEST phosphatase-interacting protein 2; pp37; proline-serine-threonine phosphatase interacting protein 2; proline-serine-threonine phosphatase-interacting protein 2; PSTPIP2; RGD1563090
Gene Symbols PSTPIP2
Host Species Rabbit
Immunogen Recombinant Human Proline-serine-threonine phosphatase-interacting protein 2 protein (28-312AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9050
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.