Learn More
BINDER™ Pt 100 Temperature Sensor
Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
BINDER Chambers KMF 115/240/720
Description
- Flexible
- Additional sensor
- Interior
Specifications
Specifications
Description | Pt 100 temperature sensor - additional flexible Pt 100, interior, for displaying the temperature on the unit display |
For Use With (Equipment) | BINDER Chambers KMF 115/240/720 |
Material | Plastic |
Type | Temperature Sensor |