Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTAFR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA525329
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA525329 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA525329 Supplier Invitrogen™ Supplier No. PA525329
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTAFR Polyclonal Antibody for Western Blot

PTAFR shows structural characteristics of the rhodopsin gene family and binds platelet-activating factor (PAF). PAF is a phospholipid (1-0-alkyl-2-acetyl-sn-glycero-3-phosphorylcholine) that has been implicated as a mediator in diverse pathologic processes, such as allergy, asthma, septic shock, arterial thrombosis, and inflammatory processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTAFR
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PTAFR
Gene Accession No. P25105, Q62035
Gene Alias PAF receptor; PAFR; PAF-R; platelet activating factor receptor; platelet-activating factor receptor; Ptafr
Gene Symbols PTAFR
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 286-312 amino acids from the C-terminal region of human PTAFR.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19204, 5724
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.