Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142305
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142305 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5142305 Supplier Invitrogen™ Supplier No. PA5142305
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognise NP_001287914.1 (isoform 1), NP_001287915.1 (isoform 2), NP_001287916.1 (isoform 3), NP_001287917.1 (isoform 4), NP_001287918.1 (isoform 5), and NP_067013.1 (isoform 6). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:1,000.

PTBP2 encodes a protein that binds to the intronic cluster of RNA regulatory elements, downstream control sequence (DCS). It is implicated in controlling the assembly of other splicing-regulatory proteins. This protein is very similar to the polypyrimidine tract binding protein but it is expressed primarily in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTBP2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene PTBP2
Gene Accession No. Q91Z31, Q9UKA9
Gene Alias br n nPTB5; brain-enriched polypyrimidine tract-binding protein; Brain-enriched PTB; Brptb; neural polypyrimidine tract binding protein; neural polypyrimidine tract-binding protein; Neurally-enriched homolog of PTB; NPTB; nPTB5; nPTB6; nPTB7; nPTB8; polypyrimidine tract binding protein 2; polypyrimidine tract-binding protein 2; PTB; Ptb2; PTB-like protein; Ptblp; PTBP2; RRM-type RNA-binding protein brPTB
Gene Symbols PTBP2
Host Species Goat
Immunogen Peptide with sequence C-SKKFKGEDKMD (aa 44-54).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56195, 58155
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.