Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTBP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541897
Description
Peptide sequence: MDGIVTEVAV GVKRGSDELL SGSVLSSPNS NMSSMVVTAN GNDSKKFKGE Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
PTBP2 encodes a protein that binds to the intronic cluster of RNA regulatory elements, downstream control sequence (DCS). It is implicated in controlling the assembly of other splicing-regulatory proteins. This protein is very similar to the polypyrimidine tract binding protein but it is expressed primarily in the brain.
Specifications
PTBP2 | |
Polyclonal | |
Unconjugated | |
PTBP2 | |
br n nPTB5; brain-enriched polypyrimidine tract-binding protein; Brain-enriched PTB; Brptb; neural polypyrimidine tract binding protein; neural polypyrimidine tract-binding protein; Neurally-enriched homolog of PTB; NPTB; nPTB5; nPTB6; nPTB7; nPTB8; polypyrimidine tract binding protein 2; polypyrimidine tract-binding protein 2; PTB; Ptb2; PTB-like protein; Ptblp; PTBP2; RRM-type RNA-binding protein brPTB | |
Rabbit | |
Protein A | |
RUO | |
58155 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UKA9 | |
PTBP2 | |
Synthetic peptide directed towards the N-terminal of human PTBP2 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction