Learn More
Invitrogen™ PTBP2 Recombinant Rabbit Monoclonal Antibody (10S9P6)

Description
Positive Samples: HeLa, RD, SH-SY5Y, Mouse testis, Rat testis Immunogen sequence: MDGIVTEVAV GVKRGSDELL SGSVLSSPNS NMSSMVVTAN GNDSKKFKGE DKMDGAPSRV LHIRKLPGEV TETEVIALGL PFGKVTNILM LKGKNQAFLE.
Specifications
Specifications
| Antigen | PTBP2 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 10S9P6 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PTBP2 |
| Gene Accession No. | Q66H20, Q91Z31, Q9UKA9 |
| Gene Alias | br n nPTB5; brain-enriched polypyrimidine tract-binding protein; Brain-enriched PTB; Brptb; neural polypyrimidine tract binding protein; neural polypyrimidine tract-binding protein; Neurally-enriched homolog of PTB; NPTB; nPTB5; nPTB6; nPTB7; nPTB8; polypyrimidine tract binding protein 2; polypyrimidine tract-binding protein 2; PTB; Ptb2; PTB-like protein; Ptblp; PTBP2; RRM-type RNA-binding protein brPTB |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.