Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTBP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565511
Description
Immunogen sequence: PAIGFPQATGL SVPAVPGALG PLTITSSAVT GRMAIPGASG IPGN Highest antigen sequence identity to the following orthologs - mouse 89%, rat 89%.
This gene is apparently required for both cerebellar and cortical development in humans. This gene mutations cause specific forms of Joubert syndrome-related disorders. Joubert syndrome (JS) is a recessively inherited developmental brain disorder with several identified causative chromosomal loci. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
PTBP3 | |
Polyclonal | |
Unconjugated | |
PTBP3 | |
5830471K22Rik; AA407443; AI462022; AW107884; C86549; fission yeast differentiation regulator; polypyrimidine tract binding protein 3; polypyrimidine tract-binding protein 3; PTBP3; regulator of differentiation (in S. pombe) 1; regulator of differentiation 1; regulator of differentiation 1; polypyrimidine tract-binding protein 3; ROD1; ROD1 regulator of differentiation 1; si:dkey-27p1.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9991 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95758 | |
PTBP3 | |
Recombinant Human PTBP3. Recombinant protein control fragment (Product #RP-106187). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction