Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTBP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598125
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598125 Supplier Invitrogen™ Supplier No. PA598125
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTBP3 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA

This gene is apparently required for both cerebellar and cortical development in humans. This gene mutations cause specific forms of Joubert syndrome-related disorders. Joubert syndrome (JS) is a recessively inherited developmental brain disorder with several identified causative chromosomal loci. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTBP3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PTBP3
Gene Accession No. O95758
Gene Alias 5830471K22Rik; AA407443; AI462022; AW107884; C86549; fission yeast differentiation regulator; polypyrimidine tract binding protein 3; polypyrimidine tract-binding protein 3; PTBP3; regulator of differentiation (in S. pombe) 1; regulator of differentiation 1; regulator of differentiation 1; polypyrimidine tract-binding protein 3; ROD1; ROD1 regulator of differentiation 1; si:dkey-27p1.2
Gene Symbols PTBP3
Host Species Rabbit
Immunogen Peptide sequence from Human Polypyrimidine tract-binding protein 3 protein (37-55AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9991
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.