Learn More
PTBP3/ROD1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305113A

Description
The recommended shelf life for this product is 1 year from date of receipt.
ROD1 is a functional homolog of nrd1, an S. pombe RNA-binding protein that suppresses the onset of differentiation.Specifications
PTBP3/ROD1 | |
Polyclonal | |
Unconjugated | |
PTBP3 | |
ROD1, ROD1 regulator of differentiation 1, fission yeast differentiation regulator, regulator of differentiation 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9991 | |
4° C | |
Liquid |
Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O95758 | |
PTBP3 | |
Between 150 and 200 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.