Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTCD2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537976
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
PTCD2 is a protein coding gene. Involved in mitochondrial RNA maturation and mitochondrial respiratory chain function.
Specifications
PTCD2 | |
Polyclonal | |
Unconjugated | |
PTCD2 | |
1190005P08Rik; CTC-365E16.1; FLJ12598; pentatricopeptide repeat domain 2; pentatricopeptide repeat-containing protein 2, mitochondrial; Ptcd2 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
79810 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8WV60 | |
PTCD2 | |
Peptide with sequence C-TLHITGQVTTDSLD, from the internal region near the C-terminus of PTCD2 (aa 338-352). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction