Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTCD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583675
Description
Immunogen sequence: KLNSPESFKI CTTLREEALL KGEILSRRAS CFAVALALNQ NEMAKAVSIF SQIMNPESIA CINLNIIIHI QSNMLENLIK TLK.
PTCD2 is a protein coding gene. Involved in mitochondrial RNA maturation and mitochondrial respiratory chain function.
Specifications
PTCD2 | |
Polyclonal | |
Unconjugated | |
PTCD2 | |
1190005P08Rik; CTC-365E16.1; FLJ12598; pentatricopeptide repeat domain 2; pentatricopeptide repeat-containing protein 2, mitochondrial; Ptcd2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79810 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WV60 | |
PTCD2 | |
Recombinant protein corresponding to Human PTCD2. Recombinant protein control fragment (Product #RP-100187). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction