Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTCD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113165
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113165 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113165 Supplier Invitrogen™ Supplier No. PA5113165
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTCD2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

PTCD2 is a protein coding gene. Involved in mitochondrial RNA maturation and mitochondrial respiratory chain function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTCD2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PTCD2
Gene Accession No. Q8R3K3, Q8WV60
Gene Alias 1190005P08Rik; CTC-365E16.1; FLJ12598; pentatricopeptide repeat domain 2; pentatricopeptide repeat-containing protein 2, mitochondrial; Ptcd2
Gene Symbols PTCD2
Host Species Rabbit
Immunogen Recombinant Human Pentatricopeptide repeat-containing protein 2, mitochondrial protein (153-388AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68927, 79810
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.