Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTCD3 Monoclonal Antibody (OTI1A8), TrueMAB™, OriGene
SDP

Catalog No. 502544026
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4026 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4026 Supplier OriGene Supplier No. CF810262
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PTCD3 Monoclonal antibody specifically detects PTCD3 in Human samples. It is validated for Western Blot

Specifications

Antigen PTCD3
Applications Western Blot
Classification Monoclonal
Clone OTI1A8
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene Mlh1
Gene Accession No. P40692
Gene Alias 2610034F17Rik; 2810422B04Rik; 28S ribosomal protein S39, mitochondrial; AA589622; AU045708; Mitochondrial small ribosomal subunit protein mS39; Mrps39; MRP-S39; pentatricopeptide repeat domain 3; pentatricopeptide repeat domain-containing protein 3, mitochondrial; pentatricopeptide repeat-containing protein 3, mitochondrial; Ptcd3; similar to CG4679-PA; transformation-related gene 15 protein; TRG15; TRG-15
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 299-527 of human MLH1 produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4292
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.