Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTCD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545963
Description
Peptide sequence: LEVIPKIWKD SKEYGHTFRS DLREEILMLM ARDKHPPELQ VAFADCAADI Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 79%; Rabbit: 93%; Rat: 86%.
Mitochondrial RNA-binding protein that has a role in mitochondrial translation.
Specifications
| PTCD3 | |
| Polyclonal | |
| Unconjugated | |
| PTCD3 | |
| 2610034F17Rik; 2810422B04Rik; 28S ribosomal protein S39, mitochondrial; AA589622; AU045708; Mitochondrial small ribosomal subunit protein mS39; Mrps39; MRP-S39; pentatricopeptide repeat domain 3; pentatricopeptide repeat domain-containing protein 3, mitochondrial; pentatricopeptide repeat-containing protein 3, mitochondrial; Ptcd3; similar to CG4679-PA; transformation-related gene 15 protein; TRG15; TRG-15 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55037 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96EY7 | |
| PTCD3 | |
| Synthetic peptide directed towards the C-terminal of human PTCD3 (aa 506-555). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction