Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTCD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597087
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597087 Supplier Invitrogen™ Supplier No. PA597087
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PDQHRNFYYS KFFDLICLME QIDVTLKWYE DLIPSAYFPH SQTMIHLLQA LDVANRLEVI PKIWKDSKEY GHTFRSDLRE EILMLMARDK HPPELQVAFA DCAADIKSAY ESQPIRQTAQ DWPATSLNCI AILFLRAGRT QEAWKMLGLF RKHNKIPRSE LLNELMDSAK VSNSPSQAIE VVELASAFSL PICEGLTQRV MSDFAINQEQ KEALSNLTAL TSDSDTDSSS DSDSDTSEGK; Positive Samples: A-549, SGC-7901, LO2, K-562; Cellular Location: Mitochondrion.

Mitochondrial RNA-binding protein that has a role in mitochondrial translation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTCD3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PTCD3
Gene Accession No. Q14C51, Q96EY7
Gene Alias 2610034F17Rik; 2810422B04Rik; 28S ribosomal protein S39, mitochondrial; AA589622; AU045708; Mitochondrial small ribosomal subunit protein mS39; Mrps39; MRP-S39; pentatricopeptide repeat domain 3; pentatricopeptide repeat domain-containing protein 3, mitochondrial; pentatricopeptide repeat-containing protein 3, mitochondrial; Ptcd3; similar to CG4679-PA; transformation-related gene 15 protein; TRG15; TRG-15
Gene Symbols PTCD3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 450-689 of human PTCD3 (NP_0604224).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 500199, 55037, 69956
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.